Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014159.6 | NP_054878.5 | 3 | 1323 | ||
XM_011533631.2 | XP_011531933.1 | 4 | 2819 | |||
XM_011533632.2 | XP_011531934.1 | 3 | 1395 | |||
XM_011533633.2 | XP_011531935.1 | 4 | 2821 | |||
XM_011533634.1 | XP_011531936.1 | 3 | 1348 | |||
XM_017006270.1 | XP_016861759.1 | 4 | 2822 | |||
GenBank mRNA | AK127782.1 | - | 1 | 740 | ||
AK128096.1 | - | 3 | 1348 | |||
AK131371.1 | - | 3 | 1313 | |||
AL831959.1 | - | 1 | 422 | |||
BC072440.1 | - | 1 | 111 | |||
BX649110.1 | - | 1 | 885 |