Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014161.4 | NP_054880.2 | 3, 4 | 616 | ||
NR_134860.1 | 4, 5 | 736 | ||||
NR_134861.1 | 3, 4 | 693 | ||||
NR_134862.1 | 3, 4 | 616 | ||||
NR_134863.1 | 3, 4 | 600 | ||||
NR_134864.1 | 3, 4 | 499 | ||||
GenBank mRNA | AF161556.1 | - | 3, 4 | 594 | ||
AK311391.1 | - | 4, 5 | 613 | |||
AL136633.1 | - | 3, 4 | 566 | |||
BC001623.1 | - | 3, 4 | 587 | 3357333 | ||
BF797811.1 | - | 3, 4 | 500 | |||
BM924328.1 | - | 3, 4 | 709 | |||
BU601065.1 | - | 3, 4 | 557 | |||
BU932963.1 | - | 3, 4 | 605 | |||
CB529016.1 | - | 1, 2 | 389 | |||
CD675657.1 | - | 3, 4 | 499 | |||
CR533491.1 | - | 3, 4 | 474 |