Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014175.3 | NP_054894.1 | 1 | 131 | ||
XM_017013362.1 | XP_016868851.1 | 1 | 142 | |||
GenBank mRNA | AF161494.1 | - | 1 | 123 | ||
AK312832.1 | - | 1 | 95 | |||
AL136665.1 | - | 1 | 101 | |||
BC000891.1 | - | 1 | 131 | 3463430 | ||
CR533531.1 | - | 1 | 68 |