Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318217.1 | NP_001305146.1 | 4, 5 | 529 | ||
NM_014187.3 | NP_054906.2 | 4, 5 | 412 | |||
GenBank mRNA | BC003080.2 | - | 4, 5 | 385 | 3050277 | |
BC013412.2 | - | 4, 5 | 364 | 3030307 | ||
BF316569.1 | - | 4, 5 | 338 | |||
BG825054.1 | - | 4, 5 | 490 |