Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001511.3 | NP_001502.1 | 3, 4 | 385 | ||
NR_046035.1 | 3, 4 | 408 | ||||
GenBank mRNA | BC011976.1 | - | 3, 4 | 373 | 3856841 | |
BG121608.1 | - | 3, 4 | 392 | |||
BM554448.1 | - | 3, 4 | 398 | |||
BT006880.1 | - | 306 | ||||
CB112830.1 | - | 3, 4 | 386 | |||
J03561.1 | - | 3, 4 | 359 | |||
X12510.1 | - | 3, 4 | 323 |