Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 7, 8 | 1001 | ||
GenBank mRNA | AK075455.1 | - | 8, 9 | 1115 | ||
AK075481.1 | - | 8, 9 | 902 | |||
AK293249.1 | - | 6, 7 | 674 | |||
BC014433.1 | - | 7, 8 | 942 | 3542405 | ||
BC036000.1 | - | 7, 8 | 955 | 4712175 | ||
BC071878.1 | - | 7, 8 | 944 | 6380973 | ||
D16234.1 | - | 7, 8 | 920 | |||
D83485.1 | - | 7, 8 | 875 | |||
EU668338.1 | - | 7, 8 | 955 | |||
FJ224330.1 | - | 7, 8 | 944 | |||
U42068.1 | - | 7, 8 | 931 | |||
Z49835.1 | - | 7, 8 | 959 |
Gene Symbol: PDIA3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002305.1 | 1 | 1116 | |||
GenBank mRNA | AK295985.1 | - | 1 | 1001 |