Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 14, 15 | 1202 | ||
NM_013314.3 | NP_037446.1 | 15, 16 | 1271 | |||
NR_047680.1 | 14, 15 | 1035 | ||||
NR_047681.1 | 13, 14 | 983 | ||||
NR_047682.1 | 13, 14 | 978 | ||||
XM_011539728.2 | XP_011538030.1 | 15, 16 | 1148 | |||
XM_017016159.1 | XP_016871648.1 | 14, 15 | 1080 | |||
GenBank mRNA | AF068180.1 | - | 15, 16 | 1246 | ||
AF068181.1 | - | 14, 15 | 1177 | |||
AK027779.1 | - | 15, 16 | 1206 | |||
AK225546.1 | - | 14, 15 | 1185 | |||
AM180333.1 | - | 14, 15 | 1024 | |||
AM180334.1 | - | 14, 15 | 1024 | |||
AM180335.1 | - | 11, 12 | 662 | |||
AM180338.1 | - | 13, 14 | 865 | |||
AM180339.1 | - | 13, 14 | 860 | |||
AM180343.1 | - | 14, 15 | 1043 | |||
AM180344.1 | - | 12, 13 | 933 | |||
AM180346.1 | - | 12, 13 | 960 | |||
BC018906.2 | - | 15, 16 | 1185 | 4053894 | ||
BC110298.1 | - | 14, 15 | 1027 | |||
BF975580.1 | - | 3, 4 | 202 |