Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014550.3 | NP_055365.2 | 10, 11 | 1668 | ||
GenBank mRNA | AK074906.1 | - | 7, 8 | 898 | ||
AY028896.1 | - | 10, 11 | 1705 | |||
AY032928.1 | - | 10, 11 | 1668 | |||
BC032058.1 | - | 8, 9 | 1293 | |||
BC113659.1 | - | 10, 11 | 1686 | 8322711 | ||
BC117403.1 | - | 10, 11 | 1680 | 40125954 |