Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199835.1 | NP_001186764.1 | 3 | 333 | ||
NM_001199837.1 | NP_001186766.1 | 1 | 138 | |||
NM_001318198.1 | NP_001305127.1 | 4 | 407 | |||
NM_001318199.1 | NP_001305128.1 | 3 | 208 | |||
NM_013322.2 | NP_037454.2 | 3 | 286 | |||
XM_006715712.2 | XP_006715775.1 | 3 | 486 | |||
XM_017012085.1 | XP_016867574.1 | 5 | 501 | |||
XM_017012086.1 | XP_016867575.1 | 5 | 940 | |||
XM_017012087.1 | XP_016867576.1 | 4 | 430 | |||
XM_017012088.1 | XP_016867577.1 | 4 | 377 | |||
GenBank mRNA | AF121860.1 | - | 3 | 199 | ||
AK296142.1 | - | 4 | 237 | |||
AK307905.1 | - | 1 | 87 | |||
AK312850.1 | - | 3 | 208 | |||
AK316088.1 | - | 4 | 383 | |||
BC031050.1 | - | 3 | 180 | 5278015 | ||
BC034992.1 | - | 3 | 259 | 4827599 | ||
BJ995940.1 | - | 1 | 138 | |||
BM147978.1 | - | 3 | 327 | |||
DA377368.1 | - | 3 | 208 |