Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039877.1 | NP_001034966.1 | 9, 10 | 1303 | ||
NM_013403.2 | NP_037535.2 | 9, 10 | 1282 | |||
XM_006723171.2 | XP_006723234.1 | 10, 11 | 1360 | |||
XM_006723172.2 | XP_006723235.1 | 10, 11 | 1339 | |||
XM_006723174.2 | XP_006723237.1 | 9, 10 | 1110 | |||
XM_011526878.1 | XP_011525180.1 | 10, 11 | 1484 | |||
XM_011526879.1 | XP_011525181.1 | 3, 4 | 447 | |||
XM_017026716.1 | XP_016882205.1 | 10, 11 | 1089 | |||
XM_017026717.1 | XP_016882206.1 | 9, 10 | 1065 | |||
XM_017026718.1 | XP_016882207.1 | 5, 6 | 854 | |||
XM_017026719.1 | XP_016882208.1 | 9, 10 | 1439 | |||
GenBank mRNA | AB209003.1 | - | 9, 10 | 1255 | ||
AF212940.1 | - | 9, 10 | 1249 | |||
AK056294.1 | - | 3, 4 | 304 | |||
AK225493.1 | - | 3, 4 | 278 | |||
AK297637.1 | - | 10, 11 | 1261 | |||
AK298804.1 | - | 10, 11 | 1329 | |||
AK308813.1 | - | 4, 5 | 517 | |||
BC004910.2 | - | 1, 2 | 37 | |||
BC034604.1 | - | 9, 10 | 1259 | 4829172 |