Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 1, 2 | 307 | ||
NM_013328.3 | NP_037460.2 | 1, 2 | 307 | |||
GenBank mRNA | AK001508.1 | - | 1, 2 | 245 | ||
AK291913.1 | - | 1, 2 | 142 | |||
BC014868.1 | - | 1, 2 | 198 | 3919161 | ||
BC020553.1 | - | 1, 2 | 232 | 2901360 | ||
BU538765.1 | - | 1, 2 | 182 | |||
HY044355.1 | - | 1, 2 | 308 | |||
KU178615.1 | - | 1, 2 | 77 | |||
KU178616.1 | - | 1, 2 | 77 |