Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013336.3 | NP_037468.1 | 12 | 1557 | ||
GenBank mRNA | AF077032.1 | - | 12 | 1463 | ||
AF084458.1 | - | 12 | 1463 | |||
AF346602.1 | - | 12 | 1439 | |||
AK001689.1 | - | 5 | 617 | |||
AK027423.1 | - | 12 | 1557 | |||
AK027452.1 | - | 4 | 523 | |||
AK027454.1 | - | 3 | 343 | |||
AK027458.1 | - | 3 | 302 | |||
AK074706.1 | - | 12 | 1557 | |||
AK074907.1 | - | 4 | 527 | |||
AK074928.1 | - | 8 | 1165 | |||
AK075148.1 | - | 4 | 557 | |||
AK129816.1 | - | 1 | 96 | |||
AK299118.1 | - | 12 | 1637 | |||
AL831940.1 | - | 7 | 967 | |||
AL832821.1 | - | 12 | 1490 | |||
BC002951.1 | - | 12 | 1464 |