Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083893.1 | NP_001077362.1 | 17 | 2414 | ||
NM_014574.3 | NP_055389.3 | 15 | 2162 | |||
XM_005267569.3 | XP_005267626.1 | 16 | 2430 | |||
XM_005267570.3 | XP_005267627.1 | 16 | 2400 | |||
GenBank mRNA | AF243424.1 | - | 16 | 1843 | ||
AK290229.1 | - | 15 | 2140 | |||
AK314041.1 | - | 15 | 2128 | |||
BC126221.1 | - | 15 | 2080 | 8991937 | ||
BC132673.1 | - | 15 | 2080 | 40146695 | ||
BC143933.1 | - | 14 | 1946 | |||
BG572065.1 | - | 170 | ||||
U17989.1 | - | 15 | 2150 |