Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 3 | 766 | ||
GenBank mRNA | AF161080.1 | - | 3 | 766 | ||
AJ400841.1 | - | 3 | 707 |