Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001092.4 | NP_001083.2 | 4 | 481 | ||
NM_001159746.2 | NP_001153218.1 | 5 | 1022 | |||
NM_001322840.1 | NP_001309769.1 | 5 | 725 | |||
NM_001322841.1 | NP_001309770.1 | 5 | 1387 | |||
NM_021962.4 | NP_068781.2 | 5 | 807 | |||
XM_011523810.2 | XP_011522112.1 | 5 | 610 | |||
XM_011523811.2 | XP_011522113.2 | 5 | 1420 | |||
XM_011523812.2 | XP_011522114.1 | 4 | 1957 | |||
XM_011523813.2 | XP_011522115.1 | 4 | 481 | |||
XM_011523814.2 | XP_011522116.1 | 5 | 1022 | |||
XM_011523815.2 | XP_011522117.1 | 5 | 705 | |||
XM_017024539.1 | XP_016880028.1 | 5 | 1417 | |||
XM_017024540.1 | XP_016880029.1 | 4 | 1319 | |||
XM_017024541.1 | XP_016880030.1 | 5 | 1420 | |||
GenBank mRNA | AK124547.1 | - | 5 | 1022 | ||
AK126882.1 | - | 5 | 609 | |||
AK295191.1 | - | 4 | 399 | |||
AK302682.1 | - | 5 | 548 | |||
AK307827.1 | - | 4 | 887 | |||
BM827672.1 | - | 4 | 465 | |||
DA106008.1 | - | 4 | 481 | |||
U01147.1 | - | 5 | 670 |