Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000014.4 | NP_000005.2 | 21, 22 | 2823 | ||
XM_006719056.2 | XP_006719119.1 | 21, 22 | 2801 | |||
GenBank mRNA | AB209614.2 | - | 21, 22 | 2734 | ||
AK307832.1 | - | 20, 21 | 2560 | |||
AK312482.1 | - | 21, 22 | 2723 | |||
AY591530.1 | - | 21, 22 | 2788 | |||
BC040071.1 | - | 21, 22 | 2762 | 6056126 | ||
BX647329.1 | - | 4, 5 | 500 | |||
BX648622.1 | - | 2207 | ||||
CR749334.1 | - | 21, 22 | 2781 | |||
M11313.1 | - | 21, 22 | 2753 | |||
M36501.1 | - | 2, 3 | 217 |