Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000180.3 | NP_000171.1 | 13 | 2540 | ||
XM_011523816.1 | XP_011522118.1 | 12 | 2466 | |||
GenBank mRNA | M92432.1 | - | 13 | 2539 |