Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005321.2 | NP_005312.1 | 1 | 222 | ||
GenBank mRNA | AK313681.1 | - | 1 | 223 | ||
BC096168.1 | - | 1 | 194 | 40003408 | ||
BC096169.1 | - | 1 | 194 | 40003411 | ||
BC096170.1 | - | 1 | 194 | 40003413 | ||
BC099632.1 | - | 1 | 194 | 40003410 | ||
EF065554.1 | - | 1 | 190 | |||
EF065555.1 | - | 1 | 190 |