Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 3 | 393 | ||
GenBank mRNA | AF117710.1 | - | 3 | 373 | ||
AK311825.1 | - | 3 | 393 | |||
AY136510.1 | - | 3 | 343 | |||
AY509193.1 | - | 3 | 393 | |||
BC007075.1 | - | 3 | 394 | 4292125 | ||
CR536530.1 | - | 3 | 343 | |||
CR541913.1 | - | 3 | 343 | |||
EU694432.1 | - | 3 | 390 | |||
M25113.1 | - | 3 | 343 | |||
V00497.1 | - | 3 | 393 | |||
V00500.1 | - | 3 | 343 |