Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 3 | 474 | ||
GenBank mRNA | AF117710.1 | - | 3 | 454 | ||
AF181832.1 | - | 3 | 367 | |||
AF181989.1 | - | 3 | 457 | |||
AF349114.1 | - | 3 | 459 | |||
AK311825.1 | - | 3 | 474 | |||
AY509193.1 | - | 3 | 474 | |||
BC007075.1 | - | 3 | 475 | 4292125 | ||
CR536530.1 | - | 3 | 424 | |||
CR541913.1 | - | 3 | 424 | |||
EU694432.1 | - | 3 | 471 | |||
M11428.1 | - | 1 | 61 | |||
M25113.1 | - | 3 | 424 | |||
V00497.1 | - | 3 | 474 | |||
V00500.1 | - | 3 | 424 |