Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002111.8 | NP_002102.4 | 32, 33 | 4394 | ||
GenBank mRNA | AB016794.1 | - | 32, 33 | 4394 | ||
AB209506.1 | - | 19, 20 | 4342 | |||
L12392.1 | - | 32, 33 | 4564 |