Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126050.1 | NP_001119522.1 | 4 | 491 | ||
NM_001126051.1 | NP_001119523.1 | 4 | 466 | |||
NM_001319186.1 | NP_001306115.1 | 4 | 512 | |||
NM_001319187.1 | NP_001306116.1 | 4 | 316 | |||
NM_001319188.1 | NP_001306117.1 | 5 | 481 | |||
NM_004494.2 | NP_004485.1 | 4 | 680 | |||
NR_135008.1 | 5 | 561 | ||||
XM_011509453.2 | XP_011507755.2 | 4 | 503 | |||
XM_011509454.2 | XP_011507756.1 | 4 | 333 | |||
XM_011509455.2 | XP_011507757.1 | 4 | 399 | |||
GenBank mRNA | AK096411.1 | - | 4 | 467 | ||
AK292325.1 | - | 4 | 316 | |||
AK298342.1 | - | 5 | 499 | |||
AK304468.1 | - | 4 | 370 | |||
AK315514.1 | - | 4 | 449 | |||
AL833592.1 | - | 6 | 773 | |||
BC018991.1 | - | 4 | 531 | 3503321 | ||
CA866443.1 | - | 5 | 395 | |||
CN392158.1 | - | 4 | 498 | |||
D16431.1 | - | 4 | 673 | |||
DA207052.1 | - | 4 | 353 | |||
DA367806.1 | - | 4 | 467 | |||
DB057555.1 | - | 5 | 481 | |||
DR159090.1 | - | 4 | 659 |