Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 6 | 1189 | ||
NM_139003.2 | NP_620572.1 | 5 | 871 | |||
NM_139004.2 | NP_620573.1 | 5 | 913 | |||
NM_139006.2 | NP_620575.1 | 6 | 1147 | |||
NM_139007.2 | NP_620576.1 | 5 | 925 | |||
NM_139008.2 | NP_620577.1 | 5 | 883 | |||
NM_139009.2 | NP_620578.1 | 6 | 1120 | |||
NM_139010.2 | NP_620579.1 | 4 | 649 | |||
NM_139011.2 | NP_620580.1 | 3 | 373 | |||
GenBank mRNA | AF079407.1 | - | 6 | 1023 | ||
AF079408.1 | - | 5 | 801 | |||
AF079409.1 | - | 5 | 759 | |||
AF115265.1 | - | 6 | 1040 | |||
AF144242.1 | - | 5 | 876 | |||
AF149804.1 | - | 5 | 755 | |||
AJ249335.1 | - | 6 | 1120 | |||
AJ249336.1 | - | 5 | 925 | |||
AJ249337.1 | - | 5 | 913 | |||
AJ249338.1 | - | 3 | 373 | |||
AJ250635.1 | - | 4 | 649 | |||
AY205604.1 | - | 5 | 790 | |||
BC074721.2 | - | 7 | 1069 | 30915171 | ||
BC117201.1 | - | 6 | 1064 | 40125752 | ||
BC117203.1 | - | 6 | 1064 | 40125754 | ||
U60319.1 | - | 6 | 1250 |