Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014589.2 | NP_055404.1 | 4 | 433 | ||
GenBank mRNA | AF189279.1 | - | 4 | 433 | ||
BC069116.1 | - | 4 | 433 |