Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 5 | 612 | ||
NM_001111113.1 | NP_001104583.1 | 3 | 459 | |||
NM_001312647.1 | NP_001299576.1 | 5 | 524 | |||
NM_001312648.1 | NP_001299577.1 | 4 | 518 | |||
NM_005339.4 | NP_005330.1 | 5 | 612 | |||
GenBank mRNA | AB022435.1 | - | 5 | 414 | ||
AB022436.1 | - | 3 | 261 | |||
AK093533.1 | - | 5 | 433 | |||
AK291454.1 | - | 3 | 364 | |||
AK307130.1 | - | 4 | 423 | |||
AK315524.1 | - | 5 | 538 | |||
BC022804.1 | - | 5 | 488 | 4893778 | ||
BC050600.1 | - | 5 | 466 | 6502901 | ||
CX786829.1 | - | 5 | 451 | |||
U58522.1 | - | 5 | 331 |