Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 4 | 359 | ||
NM_004306.3 | NP_004297.2 | 3 | 236 | |||
GenBank mRNA | AJ306450.1 | - | 4 | 335 | ||
AK223374.1 | - | 3 | 235 | |||
AK290281.1 | - | 4 | 345 | |||
BC125158.1 | - | 3 | 206 | 40120955 | ||
BG823061.1 | - | 4 | 356 | |||
CR541838.1 | - | 2 | 144 | |||
Z11502.1 | - | 3 | 223 |