Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098478.1 | NP_001091948.1 | 1, 2 | 196 | ||
NM_001098479.1 | NP_001091949.1 | 1, 2 | 196 | |||
NM_018950.2 | NP_061823.2 | 1, 2 | 196 | |||
XM_011514564.1 | XP_011512866.1 | 1, 2 | 102 | |||
XM_017010810.1 | XP_016866299.1 | 1, 2 | 102 | |||
XM_017010811.1 | XP_016866300.1 | 1, 2 | 102 | |||
XM_017010812.1 | XP_016866301.1 | 1, 2 | 102 | |||
XM_017010813.1 | XP_016866302.1 | 1, 2 | 102 | |||
XM_017010814.1 | XP_016866303.1 | 1, 2 | 83 | |||
GenBank mRNA | AK096962.1 | - | 1, 2 | 102 | ||
AK125274.1 | - | 1, 2 | 106 | |||
AK298704.1 | - | 1, 2 | 102 | |||
AK302420.1 | - | 1, 2 | 135 | |||
AK309156.1 | - | 1, 2 | 102 | |||
AY221102.1 | - | 1, 2 | 60 | |||
AY253270.1 | - | 1, 2 | 72 | |||
AY253271.1 | - | 1, 2 | 72 | |||
CT002787.1 | - | 1, 2 | 115 | |||
DQ367723.1 | - | 1, 2 | 72 | |||
GQ891552.1 | - | 1, 2 | 72 |