Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098478.1 | NP_001091948.1 | 3 | 645 | ||
NM_001098479.1 | NP_001091949.1 | 3 | 645 | |||
NM_018950.2 | NP_061823.2 | 3 | 645 | |||
XM_011514564.1 | XP_011512866.1 | 3 | 551 | |||
XM_017010810.1 | XP_016866299.1 | 3 | 551 | |||
XM_017010811.1 | XP_016866300.1 | 3 | 551 | |||
XM_017010812.1 | XP_016866301.1 | 3 | 551 | |||
XM_017010813.1 | XP_016866302.1 | 3 | 551 | |||
XM_017010814.1 | XP_016866303.1 | 3 | 515 | |||
XM_017010815.1 | XP_016866304.1 | 2 | 440 | |||
GenBank mRNA | AK096962.1 | - | 3 | 551 | ||
AK225185.1 | - | 3 | 549 | |||
AK298704.1 | - | 3 | 551 | |||
AK300090.1 | - | 3 | 584 | |||
AK301488.1 | - | 3 | 532 | |||
AK302420.1 | - | 4 | 515 | |||
AK303670.1 | - | 2 | 607 | |||
AK309156.1 | - | 3 | 551 | |||
AY216682.1 | - | 3 | 509 | |||
AY221102.1 | - | 3 | 509 | |||
AY253269.1 | - | 3 | 521 | |||
AY253270.1 | - | 3 | 521 | |||
AY253271.1 | - | 3 | 521 | |||
BC009260.2 | - | 3 | 577 | |||
BC062991.1 | - | 3 | 525 | 5496696 | ||
CB528356.1 | - | 5 | 647 | |||
DQ367723.1 | - | 3 | 521 | |||
GQ891552.1 | - | 3 | 521 |