Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005519.1 | NP_005510.1 | 2 | 577 | ||
XM_005269743.3 | XP_005269800.1 | 3 | 439 | |||
GenBank mRNA | BC132758.1 | - | 2 | 387 | 40146780 | |
BC137139.1 | - | 2 | 387 | 9021136 | ||
BQ929484.1 | - | 2 | 577 |