Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152467.3 | NP_689680.2 | 3, 4 | 1435 | ||
XM_011524706.2 | XP_011523008.1 | 5, 6 | 1568 | |||
GenBank mRNA | AK057224.1 | - | 3, 4 | 1398 | ||
AK301797.1 | - | 3, 4 | 1339 | |||
AY495339.1 | - | 3, 4 | 1417 | |||
BC067753.1 | - | 3, 4 | 1422 |