Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPH3 SIRNA Assays ›
Gene Symbol:
HNRNPH3
Gene Name:
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases:
2H9, HNRPH3
Chromosome Location:
Chr.10: 68331174 - 68343196 on Build GRCh38
UniGene ID:
Hs.643472
Species:
Human
siRNA ID
HSS179310
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPH3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322434.1
NP_001309363.1
4
525
NM_001322436.1
NP_001309365.1
4
487
NM_001322437.1
NP_001309366.1
4
590
NM_001322438.1
NP_001309367.1
4
462
NM_001322439.1
NP_001309368.1
4
525
NM_001322440.1
NP_001309369.1
4
487
NM_001322441.1
NP_001309370.1
4
590
NM_001322442.1
NP_001309371.1
4
525
NM_001322443.1
NP_001309372.1
4
506
NM_001322445.1
NP_001309374.1
3
323
NM_001322446.1
NP_001309375.1
3
367
NM_001322447.1
NP_001309376.1
3
348
NM_001322448.1
NP_001309377.1
3
386
NM_001322449.1
NP_001309378.1
3
451
NM_001322450.1
NP_001309379.1
3
386
NM_001322451.1
NP_001309380.1
3
348
NM_001322452.1
NP_001309381.1
3
367
NM_001322453.1
NP_001309382.1
3
386
NM_012207.2
NP_036339.1
4
506
NM_021644.3
NP_067676.2
4
506
XM_017016168.1
XP_016871657.1
4
936
XM_017016169.1
XP_016871658.1
3
797
GenBank mRNA
AF132361.1
-
4
395
AF132362.1
-
2
407
AF132363.1
-
1
189
AK091411.1
-
2
2378
AK223441.1
-
4
458
AK291547.1
-
4
458
AL117395.1
-
1
1728
BC039824.1
-
4
442
6061550
DA486117.1
-
3
367
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA processing
RNA splicing
epithelial cell differentiation
nucleotide binding
RNA binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0