Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318186.1 | NP_001305115.1 | 5, 6 | 440 | ||
NM_001318187.1 | NP_001305116.1 | 5, 6 | 440 | |||
NM_001318188.1 | NP_001305117.1 | 5, 6 | 440 | |||
NM_002140.4 | NP_002131.2 | 5, 6 | 601 | |||
NM_031262.3 | NP_112552.1 | 5, 6 | 601 | |||
NM_031263.3 | NP_112553.1 | 5, 6 | 440 | |||
XM_005251960.2 | XP_005252017.1 | 5, 6 | 419 | |||
XM_005251963.3 | XP_005252020.1 | 5, 6 | 453 | |||
XM_005251965.2 | XP_005252022.1 | 5, 6 | 411 | |||
XM_011518616.1 | XP_011516918.1 | 5, 6 | 453 | |||
XM_017014668.1 | XP_016870157.1 | 5, 6 | 419 | |||
XM_017014669.1 | XP_016870158.1 | 5, 6 | 411 | |||
GenBank mRNA | AB209562.1 | - | 5, 6 | 369 | ||
AB451263.1 | - | 3, 4 | 214 | |||
AB451390.1 | - | 3, 4 | 214 | |||
AK096385.1 | - | 5, 6 | 298 | |||
AK291336.1 | - | 5, 6 | 423 | |||
AK300744.1 | - | 5, 6 | 383 | |||
AK307962.1 | - | 5, 6 | 331 | |||
AY911506.1 | - | 4, 5 | 277 | |||
BC000355.2 | - | 4, 5 | 336 | 2964383 | ||
BC014980.1 | - | 5, 6 | 407 | 4906241 | ||
BC025321.1 | - | 5, 6 | 359 | |||
CR456771.1 | - | 3, 4 | 214 | |||
DA394419.1 | - | 5, 6 | 438 | |||
DC318847.1 | - | 5, 6 | 440 | |||
DC407458.1 | - | 5, 6 | 473 | |||
S74678.1 | - | 4, 5 | 321 | |||
X72727.1 | - | 5, 6 | 423 |