Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005335.1 | NP_001005335.1 | 3 | 353 | ||
NM_001533.2 | NP_001524.2 | 3 | 587 | |||
XM_011526886.1 | XP_011525188.1 | 3 | 376 | |||
XM_011526887.1 | XP_011525189.1 | 3 | 420 | |||
XM_011526888.2 | XP_011525190.1 | 3 | 658 | |||
XM_011526889.1 | XP_011525191.1 | 3 | 471 | |||
GenBank mRNA | AB044547.1 | - | 3 | 582 | ||
AK292115.1 | - | 3 | 352 | |||
AK298384.1 | - | 3 | 423 | |||
AK301062.1 | - | 3 | 361 | |||
AK313652.1 | - | 3 | 522 | |||
BC069184.1 | - | 3 | 526 | |||
BI256815.1 | - | 3 | 353 | |||
X16135.1 | - | 3 | 511 |