Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130442.2 | NP_001123914.1 | 4 | 512 | ||
NM_001318054.1 | NP_001304983.1 | 4 | 512 | |||
NM_005343.3 | NP_005334.1 | 4 | 512 | |||
NM_176795.4 | NP_789765.1 | 4 | 512 | |||
GenBank mRNA | AB451336.1 | - | 3 | 307 | ||
AB451485.1 | - | 3 | 307 | |||
AF493916.1 | - | 3 | 307 | |||
AJ437024.1 | - | 3 | 307 | |||
BC006499.2 | - | 4 | 483 | 2819996 | ||
BC095471.1 | - | 3 | 308 | 6519193 | ||
BM801600.1 | - | 4 | 435 | |||
BM801632.1 | - | 4 | 495 | |||
BM808879.1 | - | 520 | ||||
BQ674260.1 | - | 3 | 370 | |||
BT019421.1 | - | 3 | 307 | |||
CR536579.1 | - | 3 | 307 | |||
CR542271.1 | - | 3 | 307 | |||
KJ534867.1 | - | 3 | 307 | |||
KJ534868.1 | - | 3 | 307 |