Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000198.3 | NP_000189.1 | 4 | 679 | ||
NM_001166120.1 | NP_001159592.1 | 4 | 756 | |||
GenBank mRNA | AK222997.1 | - | 4 | 677 | ||
AK313413.1 | - | 4 | 649 | |||
BC038419.1 | - | 4 | 654 | 5167092 | ||
BC131488.1 | - | 3 | 1281 | 40037863 | ||
M67466.1 | - | 4 | 640 |