Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACACB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093.3 | NP_001084.3 | 14 | 2380 | ||
XM_005253876.4 | XP_005253933.1 | 15 | 2490 | |||
XM_006719367.3 | XP_006719430.1 | 14 | 2011 | |||
XM_011538259.2 | XP_011536561.1 | 15 | 3128 | |||
XM_011538263.2 | XP_011536565.1 | 15 | 3128 | |||
XM_011538264.2 | XP_011536566.1 | 14 | 2465 | |||
XM_011538265.2 | XP_011536567.1 | 15 | 3130 | |||
XM_017019252.1 | XP_016874741.1 | 14 | 1995 | |||
GenBank mRNA | AB209110.1 | - | 4 | 434 | ||
AJ575431.2 | - | 14 | 2371 | |||
AJ575592.3 | - | 14 | 2371 | |||
AK307466.1 | - | 9 | 1233 | |||
AY382667.1 | - | 15 | 2490 | |||
DQ493870.1 | - | 14 | 2371 | |||
U89344.1 | - | 14 | 2368 |