Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 12 | 1741 | ||
NM_199440.1 | NP_955472.1 | 12 | 1721 | |||
GenBank mRNA | AK297305.1 | - | 1668 | |||
AK298632.1 | - | 12 | 1664 | |||
AK298844.1 | - | 11 | 1492 | |||
AK312240.1 | - | 12 | 1674 | |||
BC002676.2 | - | 12 | 1651 | 3609039 | ||
BC003030.1 | - | 12 | 1650 | 2821157 | ||
BC047350.1 | - | 11 | 1811 | |||
BC067082.1 | - | 12 | 1649 | 4025725 | ||
BC073746.1 | - | 12 | 1649 | 6372068 | ||
BX647145.1 | - | 11 | 2406 | |||
EU719626.1 | - | 11 | 1604 | |||
KU178139.1 | - | 11 | 1604 | |||
KU178140.1 | - | 6 | 855 | |||
KU178141.1 | - | 3 | 462 | |||
M22382.1 | - | 12 | 1649 | |||
M34664.1 | - | 12 | 1628 |