Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 12 | 1843 | ||
NM_199440.1 | NP_955472.1 | 12 | 1823 | |||
GenBank mRNA | AK297305.1 | - | 1770 | |||
AK298632.1 | - | 12 | 1766 | |||
AK298844.1 | - | 11 | 1594 | |||
AK312240.1 | - | 12 | 1776 | |||
BC002676.2 | - | 12 | 1753 | 3609039 | ||
BC003030.1 | - | 12 | 1752 | 2821157 | ||
BC047350.1 | - | 11 | 1913 | |||
BC067082.1 | - | 12 | 1751 | 4025725 | ||
BC073746.1 | - | 12 | 1751 | 6372068 | ||
BX647145.1 | - | 11 | 2508 | |||
M22382.1 | - | 12 | 1751 | |||
M34664.1 | - | 12 | 1730 |