Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291860.1 | NP_001278789.1 | 67 | 8871 | ||
NM_005529.6 | NP_005520.4 | 67 | 8868 | |||
XM_011541318.2 | XP_011539620.1 | 72 | 9352 | |||
XM_017001120.1 | XP_016856609.1 | 68 | 8998 | |||
XM_017001121.1 | XP_016856610.1 | 67 | 8947 | |||
XM_017001122.1 | XP_016856611.1 | 67 | 8944 | |||
GenBank mRNA | AB209851.1 | - | 19 | 2608 | ||
M85289.1 | - | 67 | 8868 | |||
X62515.1 | - | 67 | 8834 |