Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000866.4 | NP_000857.1 | 2 | 694 | ||
NM_001322208.1 | NP_001309137.1 | 3 | 975 | |||
NM_001322209.1 | NP_001309138.1 | 3 | 771 | |||
NM_001322210.1 | NP_001309139.1 | 3 | 762 | |||
XM_005264751.3 | XP_005264808.1 | 3 | 841 | |||
XM_011533664.2 | XP_011531966.1 | 3 | 773 | |||
GenBank mRNA | AF498981.1 | - | 1 | 536 | ||
BC069125.1 | - | 1 | 536 | 7216980 |
Set Membership: |
G-protein coupled receptor |