Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000871.2 | NP_000862.1 | 2 | 1299 | ||
GenBank mRNA | AB209129.1 | - | 1 | 5758 | ||
AY429105.1 | - | 2 | 832 | |||
BC074995.2 | - | 2 | 866 | 30915396 | ||
BC074996.2 | - | 2 | 866 | 30915608 | ||
L41147.1 | - | 2 | 1299 |
Set Membership: |
G-protein coupled receptor |