Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002161.5 | NP_002152.2 | 30 | 3395 | ||
NM_013417.3 | NP_038203.2 | 30 | 3565 | |||
NR_073446.1 | 29 | 3238 | ||||
GenBank mRNA | AB209234.1 | - | 30 | 3304 | ||
AK057920.1 | - | 2 | 1140 | |||
AK293014.1 | - | 30 | 3322 | |||
AK301272.1 | - | 18 | 1875 | |||
AK308202.1 | - | 11 | 1132 | |||
BC008318.1 | - | 8 | 900 | |||
BC065552.1 | - | 29 | 3237 | 6043344 | ||
BX537417.1 | - | 30 | 3333 | |||
BX537429.1 | - | 30 | 3335 | |||
BX538109.1 | - | 29 | 3178 | |||
BX647071.1 | - | 30 | 3334 | |||
BX647236.1 | - | 30 | 3327 | |||
BX647430.1 | - | 30 | 3329 | |||
BX648567.1 | - | 30 | 3335 | |||
D28473.1 | - | 30 | 3483 | |||
U04953.1 | - | 30 | 3497 |