Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198489.2 | NP_940891.1 | 3 | 367 | ||
NR_104049.1 | 3 | 367 | ||||
NR_104050.1 | 3 | 367 | ||||
NR_104051.1 | 3 | 367 | ||||
XM_011542799.1 | XP_011541101.1 | 2 | 182 | |||
XM_011542800.2 | XP_011541102.1 | 1 | 18 | |||
XM_017017652.1 | XP_016873141.1 | 3 | 433 | |||
GenBank mRNA | AB094093.1 | - | 3 | 358 | ||
BC126469.1 | - | 3 | 332 | 8992185 | ||
BC126471.1 | - | 3 | 332 | 8992187 | ||
BC144208.1 | - | 3 | 332 | |||
BC144209.1 | - | 3 | 332 | |||
BC144210.1 | - | 3 | 332 | |||
HY073607.1 | - | 3 | 369 |