Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178543.4 | NP_848638.3 | 3 | 889 | ||
XM_011524737.1 | XP_011523039.1 | 3 | 1073 | |||
GenBank mRNA | AK126250.1 | - | 3 | 1073 | ||
AY230663.1 | - | 3 | 872 | |||
AY358622.1 | - | 3 | 855 | |||
BC041453.1 | - | 3 | 872 | 5186743 |