Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012241.1 | NP_001012241.1 | 4, 5 | 936 | ||
XM_005257298.4 | XP_005257355.1 | 3, 4 | 2588 | |||
GenBank mRNA | AK297619.1 | - | 1, 2 | 257 | ||
BC118997.1 | - | 4, 5 | 859 | 40084759 | ||
BC122543.1 | - | 4, 5 | 859 | 40084757 | ||
CR749360.1 | - | 4, 5 | 887 |