Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001664.2 | NP_001001664.1 | 3 | 543 | ||
XM_005263655.4 | XP_005263712.1 | 3 | 409 | |||
XM_005263656.4 | XP_005263713.1 | 3 | 409 | |||
XM_005263657.4 | XP_005263714.1 | 3 | 419 | |||
GenBank mRNA | BC071613.1 | - | 3 | 522 | 5275316 |