Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002167.4 | NP_002158.3 | 2 | 694 | ||
GenBank mRNA | A17546.1 | - | 2 | 449 | ||
A17548.1 | - | 2 | 326 | |||
AK225965.1 | - | 1 | 536 | |||
AK290003.1 | - | 2 | 431 | |||
BC003107.1 | - | 2 | 412 | 3543936 | ||
BM921819.1 | - | 725 | ||||
BT006791.1 | - | 2 | 326 | |||
X66924.1 | - | 2 | 449 | |||
X69111.1 | - | 2 | 693 |