Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289910.1 | NP_001276839.1 | 6, 7 | 740 | ||
NM_001290114.1 | NP_001277043.1 | 4, 5 | 715 | |||
NM_002168.3 | NP_002159.2 | 6, 7 | 973 | |||
GenBank mRNA | AK294148.1 | - | 6, 7 | 740 | ||
AK299987.1 | - | 4, 5 | 629 | |||
AK312627.1 | - | 6, 7 | 937 | |||
AK316388.1 | - | 6, 7 | 814 | |||
BC009244.2 | - | 6, 7 | 875 | 2959540 | ||
BC071828.1 | - | 6, 7 | 874 | 6090792 | ||
X69433.1 | - | 6, 7 | 895 |