Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080532.1 | NP_001074001.1 | 12, 13 | 1273 | ||
GenBank mRNA | AY236490.1 | - | 12, 13 | 1351 | ||
AY263163.1 | - | 12, 13 | 1312 | |||
BC132679.1 | - | 12, 13 | 1310 | 40146701 | ||
BC143410.1 | - | 12, 13 | 1310 | 9051917 | ||
BC157874.1 | - | 12, 13 | 1322 | 9057092 |