Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010985.2 | NP_001010985.2 | 1 | 89 | ||
NM_001265613.1 | NP_001252542.1 | 1 | 89 | |||
XM_017001173.1 | XP_016856662.1 | 1 | 2442 | |||
XM_017001174.1 | XP_016856663.1 | 1 | 2442 | |||
XM_017001175.1 | XP_016856664.1 | 1 | 2442 | |||
GenBank mRNA | AK129834.1 | - | 1 | 89 | ||
BC108664.1 | - | 1 | 44 | |||
BC132900.1 | - | 1 | 66 | 40146922 | ||
BC132902.1 | - | 1 | 66 | 40146924 | ||
BC144471.1 | - | 1 | 66 | 9052998 |